Learn More
MTP Mouse, Unlabeled, Clone: 8, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 612022
Description
The microsomal triglyceride transfer protein (MTP) catalyzes the transport of triglyceride, cholesteryl ester, and phospholipid between membranes within the lumen of microsomes in hepatocytes and enterocytes. MTP forms a heterodimer with the 58kDa protein disulfide isomerase. PDI catalyzes the isomerization of intramolecular disulfide bridges, thereby allowing them to generate their most thermodynamically stable configuration within proteins. MTP is mutated in abetalipoproteinemia, which results from defects in apolipoprotein-B (apoB)-containing lipoproteins. A lack of MTP expression prevents secretion of apoB from mammalian cells, leading to intracellular degradation. In the C-terminal region, MTP has structural homology to apoB and the lamprey lipovitellin protein. This region contains a membrane binding helix (Helix A), and a triglyceride binding helix (Helix B). Mutations in Helix B cause abetalipoproteinemia. In addition, inhibitors of MTP activity may be important therapeutics for lowering atherogenic lipoprotein levels. Thus, MTP is a microsomal protein that is required for transport of lipids between membranes in liver and small intestines.
Host Species: Mouse
Clone: 8
Isotype: IgG2a
Species Reactivity [for Features Main]: Mouse
Immunogen: Mouse MTP aa. 91-288
Immunofluorescence, Western Blotting

Specifications
MTP | |
Monoclonal | |
250μg/mL | |
Microsomal Triglyceride transfer Protein | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
Western Blot | |
8 | |
Unconjugated | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Mouse MTP aa. 91-288 | |
50 μg | |
Cell Biology | |
Mouse, Rat | |
IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.