Learn More
anti-MTSS1, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP224717SS
 View more versions of this product
Catalog No. NBP224717SS
Add to cart
Description
Description
MTSS1 Polyclonal antibody specifically detects MTSS1 in Human, Mouse, Rat, Canine, Chicken, Primate samples. It is validated for Western Blot.
Specifications
Specifications
MTSS1
0.5 mg/mL
Western Blot 2-4 ug/ml
O43312
FLJ44694, KIAA0429Missing in metastasis protein, metastasis suppressor 1, metastasis suppressor protein 1, Metastasis suppressor YGL-1, MIMA, MIMB, MIMDKFZp781P2223, MTSS1
Rabbit
IgG
0.025 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_c, CRA_d, CRA_e, CRA_f, CRA_g) and chimpanzee, 94% homologous in monkey and 82% homologous in dog.
Western Blot
Unconjugated
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Purified
MTSS1
A portion of amino acid 150-200 of human MTSS1 protein was used as the immunogen.
Protein G purified
RUO
Primary
9788
Human, Mouse, Rat, Canine, Chicken, Primate