Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MTTP Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA526077
Description
This antibody is predicted to react with mouse based on sequence homology.
MTTP encodes the large subunit of the heterodimeric microsomal triglyceride transfer protein. Protein disulfide isomerase (PDI) completes the heterodimeric microsomal triglyceride transfer protein, which has been shown to play a entral role in lipoprotein assembly. Mutations in MTP can cause abetalipoproteinemia.Specifications
MTTP | |
Polyclonal | |
MTTP | |
ABL, MTP, microsomal triglyceride transfer protein (large polypeptide, 88kDa), microsomal triglyceride transfer protein large subunit | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4547 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
Unconjugated | |
P55157 | |
MTTP | |
KLH conjugated synthetic peptide between 816-842 amino acids from the C-terminal region of human MTTP | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction