Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-MUC-4, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications

Manufacturer:  Novus Biologicals NBP152193

 View more versions of this product

Catalog No. NBP152193


Add to cart

Description

Description

MUC4 Polyclonal specifically detects MUC4 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

MUC-4
0.56 mg/mL
Western Blot, Simple Western 1:500, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:250, Immunohistochemistry-Paraffin 1:400
Q99102, Q8JZM8
Ascites sialoglycoprotein, ASGP, HSA276359, MUC-4, mucin 4, cell surface associated, mucin 4, tracheobronchial, mucin-4, Pancreatic adenocarcinoma mucin, Testis mucin, Tracheobronchial mucin
Rabbit
IgG
0.1 ml
Cancer
Primary
4585
Human, Mouse
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
PBS, 0.1% BSA, 50% Glycerol with 0.05% Sodium Azide
Affinity Purified
MUC4
A genomic peptide made to an internal region of the human MUC4 protein (within residues 1950-2100). [Swiss-Prot Q99102]
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
This antibody is specific for the beta chain of MUC4.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only