Learn More
MUC1 Mouse anti-Human, Clone: VU-5F12, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA135700
Catalog No. 01-672-006
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
The MA1-35700 immunogen is from a MUC1 60mer tandem repeat NH2-(VTSAPDTRPAPGSTAPPAHG)3-COOH conjugated to BSA.
MUC1 (Mucin 1, Episialin, MAM-6, CA 15-3, PEM and EMA) is a large cell surface mucin glycoprotein expressed by most glandular and ductal epithelial cells and some hematopoietic cell lineages. MUC1 is a transmembrane glycoprotein with a large mucin-like extracellular domain that matures through several intermediate forms generated by proteolysis, and sequential addition and processing of numerous O-linked glycans that are heavily sialylated. MUC1 is highly polymorphic and each allele encodes a product that contains a different number of repeats (between 30 and 90) leading to large differences in molecular weight of the protein. MUC1 is expressed on most secretory epithelium, including mammary gland and some hematopoietic cells, and is expressed abundantly in >90% breast carcinomas and metastases. Transgenic MUC1 has been shown to associate with all four cebB receptors and localize with erbB1 (EGFR) in lactating glands. The MUC1 gene contains seven exons and produces several different alternatively spliced variants. Overexpression, aberrant intracellular localization, and changes in glycosylation of the MUC1 protein have been associated with carcinomas. Multiple alternatively spliced transcript variants that encode different isoforms of the MUC1 gene have been reported, but the full-length nature of only some has been determined. Transgenic MUC-1 has been shown to associate with all four cebB receptors and localize with erbB1 (EGFR) in lactating glands.
Specifications
Specifications
MUC1
Monoclonal
Unconjugated
MUC1
Liquid
MUC1
MUC1 conjugated to BSA.
Purified
RUO
Antibody
Monoclonal
Human
ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
VU-5F12
PBS with 0.05% sodium azide
P15941
Epithelial Membrane Antigen, EMA, CD227, Episialin, CD227, EMA, H23AG, KL-6, MAM6, MUC-1, MUC-1/SEC, MUC-1/X, MUC1/ZD, PEM, PEMT, PUM, DF3 antigen, H23 antigen, breast carcinoma-associated antigen DF3
Mouse
IgG1
100 μg
4° C
Primary
4582