Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC12 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA14175

Catalog No. PA14175


Add to cart

Description

Description

PA1-4175 detects MUC12 in human samples. PA1-4175 has been successfully used in Western blot procedures. The PA1-4175 immunogen is a synthetic peptide corresponding to residues S(10) S I S G E P T S L Y S Q A E(24) of human MUC12.

Epitheial mucins are secreted glycoproteins that play roles in cell protection, adhesion modulation and signaling. MUC12 is a novel mucin gene and may be involved in epithelial cell growth regulation. MUC12 is down-regulated in colorectal cancers.
Specifications

Specifications

MUC12
Polyclonal
Unconjugated
MUC12
Liquid
MUC12
Synthetic peptide corresponding to residues S(10) S I S G E P T S L Y S Q A E(24) of human MUC12.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q9UKN1
Mucin12, Transmembrane mucin 12
Rabbit
IgG
100 μg
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Primary
10071
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit