Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC16, Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of MUC16.

Manufacturer:  Abnova Corporation PAB17423

Catalog No. 89-024-838


Add to cart

Specifications

Specifications

MUC16
Unconjugated
Immunofluorescence (1:500-1:1000) ELISA (1:40000) The optimal working dilution should be determined by the end user.
MUC16
CA125|FLJ14303
Rabbit
100 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
This antibody detects endogenous levels of total MUC16 protein.
ELISA, Immunofluorescence, Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of MUC16.
In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol)
Liquid
MUC16
A synthetic peptide corresponding to internal of human MUC16.
RUO
Primary
94025
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit