Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUPP1 Mouse, Unlabeled, Clone: 43, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611559

 View more versions of this product

Catalog No. BDB611559


Add to cart

Description

Description

The postsynaptic density protein PSD-95 interacts with the cytoplasmic tail of ion channel subunits via a protein-protein motif called the PDZ domain. PSD-95 belongs to a family of PDZ domain-containing proteins that includes SAP97/Dlg, PSD-93/chapsyn-110, and SAP102. These proteins are characterized by three PDZ domains in the N-terminal half and an SH3 and guanylate kinase-like domain in the C-terminal region. They may be involved in the localization and clustering of ion channels and receptors at synaptic sites. Other PDZ domain-containing proteins such as glutamate receptor interacting protein (GRIP), Homer, and multi-PDZ-domain protein (MUPP1) do not contain guanylate kinase-like domains, but have also been implicated in receptor localization. MUPP1 contains thirteen PDZ domains and is expressed in human heart, brain, placenta, liver, skeletal muscle, kidney, and pancreas. In addition, smaller variants of MUPP1 are expressed in heart, liver, kidney, and brain. The C-terminal region of the 5-HT2c receptor interacts with MUPP1, suggesting that MUPP1 may have important protein-protein interactions during G-protein coupled receptor signaling.

Host Species: Mouse
Clone: 43
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Rat MUPP1 aa. 65-247
Formula Weight [Chemical]: 219kDa

Immunofluorescence, Western Blotting

Specifications

Specifications

MUPP1
43
Unconjugated
Affinity Purified
Rat MUPP1 aa. 65-247
150μg
Cell Biology
Primary
Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.