Learn More
MURF1, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA522112
Catalog No. PIPA522112
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
PA5-22112 targets MURF1 in ICC, IF, IHC (P), and WB applications and shows reactivity with Human samples.The PA5-22112 immunogen is synthetic peptide contain a sequence corresponding to a region within amino acids 1 and 44 (Q969Q1) of MURF1.
Specifications
Specifications
MURF1
Polyclonal
Unconjugated
TRIM63
Liquid
TRIM63
Synthetic peptide corresponding to a region within amino acids 1 and 44 of MURF1 (Uniprot ID#Q969Q1)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
2.25mg/mL
PBS with 20% glycerol and 0.025% Proclin 300; pH 7
Q969Q1, Q38HM4
IRF, MURF1, MURF2, RNF28, SMRZ, E3 ubiquitin-protein ligase TRIM63, iris ring finger protein, muscle specific ring finger protein 2, muscle-specific RING finger protein 1, ring finger protein 28, striated muscle RING zinc finger protein, tripartite motif-containing 63, tripartite motif-containing protein 63
Rabbit
IgG
100μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
433766, 84676