Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUS81 Mouse anti-Human, Clone: 2G10/3, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA15837

Catalog No. 01-674-940


Add to cart

Description

Description

MA1-5837 detects Mus81 in human samples. MA1-5837 has been successfully used in Western blot and immunoprecipitation procedures. The MA1-5837 immunogen is human Mus81.

Interacts with EME1 and EME2 to form a DNA structure-specific endonuclease with substrate preference for branched DNA structures with a 5 ft.-end at the branch nick. Typical substrates include 3 ft.-flap structures, replication forks and nicked Holliday junctions. May be required in mitosis for the processing of stalled or collapsed replication forks.
Specifications

Specifications

Mus81
Monoclonal
1 mg/mL
PBS with 0.1% sodium azide
Q96NY9
FLJ21012, FLJ44872 MUS81
Mouse
IgG2a
50 μL
Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Primary
80198
Immunoprecipitation, Western Blot
2G10/3
Unconjugated
MUS81
Liquid
MUS81
Human Mus81 from Sp2/0 Ag.14 cells
Protein A
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit