Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Mus81, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP132054

Catalog No. NBP132054

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Mus81
2.0mg/mL
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
Purified
MUS81
Recombinant protein encompassing a sequence within the center region of human MUS81. The exact sequence is proprietary.
61 kDa
0.1 ml
DNA Repair, Homologous Recombination
Primary
80198
Human, Mouse, Rat
Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
PBS (pH 7.0) and 40% Glycerol with 0.01% Thimerosal
crossover junction endonuclease MUS81, EC 3.1.22, EC 3.1.22.-, FLJ21012, FLJ44872, MUS81 endonuclease homolog (S. cerevisiae), MUS81 endonuclease homolog (yeast), SLX3, SLX3 structure-specific endonuclease subunit homolog
Rabbit
IgG
Protein A purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in Human, Mouse and Rat. Not yet tested in any other species.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only