Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Musashi-1, Clone: EP1302, ™ Non-distribution product as customer accommodation.

Manufacturer:  MilliporeSigma™ 041041/DEL

Catalog No. 50-171-167

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

In mammals, the Musashi family is important for cell fate determination, playing roles in maintenance of the stem-cell state, differentiation and tumorigenesis. Musashi-1 (also known as Msi1), is selectively expressed in neural progenitor cells, including neural stem cells. Outside the nervous system, Musashi-1 is a selective marker for intestinal stem or early lineage cells. Musashi-1 interacts with the Notch pathway during asymmetric cell division by binding the 3' UTR of Numb. Numb is prevented from repressing Notch signaling when Musashi-1 is present.

Host Species: Rabbit
Clone: EP1302
Isotype: IgG
Species Reactivity: Human, Rat
Epitope: N-terminus
Immunogen: Synthetic peptide corresponding to residues on the N-terminus of human Musashi-1.
Formula Weight [Chemical]: ~39kDa

Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting

Specifications

Specifications

Musashi-1
Unconjugated
Rabbit
IgG
RUO
Primary
Human, Rat
EP1302
Unpurified
Synthetic peptide corresponding to residues on the N-terminus of human Musashi-1.
100μL
Neuroscience
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit