Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSI1 Rabbit anti-Canine, Equine, Human, Mouse, Non-human primate, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523439

Catalog No. PIPA523439


Add to cart

Description

Description

Musashi-1 is a member of an evolutionarily conserved family of RNA-binding proteins that is prefentially expressed in the nervous system. It is a neural RNA-binding protein that is strongly expressed in fetal and adult neural stem cells (NSCs). Functionally, Musashi-1 plays important roles in cell fate decision, including the maintece of the stem-cell state, differentiation and tumorigenesis. Musashi-1 is an important regulator of the Notch-1 pathway.
Specifications

Specifications

MSI1
Polyclonal
Unconjugated
MSI1
Liquid
LOC100053469, MSI1
Synthetic peptide (233 TGLAPGYTYQFPEFRVER 250) of Musashi-1 protein.
Protein A
RUO
Antibody
Polyclonal
Canine, Equine, Human, Mouse, Non-human primate, Rat
Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
O43347, Q61474, Q8K3P4
RNA-binding protein Musashi homolog 1, musashi-1, musashi1
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
100053469, 17690, 259272, 4440, 611488
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit