Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUTYH Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526167

Catalog No. PIPA526167

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

MYH is the eukaryotic homolog to the E. coli protein MutY. It was first identified in HeLa cells and nicks DNA replication errors, specifically A/G mismatches. MYH is involved in oxidative DNA damage repair. It initiates repair of A*oxoG to C*G by removing the inappropriately paired adenine base from the DNA backbone. It possesses both adenine and 2-OH-A DNA glycosylase activities.
Specifications

Specifications

MUTYH
Polyclonal
PBS with 0.09% sodium azide
Q9UIF7, Q99P21
CYP2C, MYH, A/G-specific adenine DNA glycosylase
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4595, 70603
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
MUTYH
Liquid
MUTYH
KLH conjugated synthetic peptide between 341-367 amino acids from the central region of human MUTYH
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit