Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mevalonate (diphospho) decarboxylase (B02), Mouse anti-Human, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human MVD protein.

Manufacturer:  Abnova Corporation H00004597B02

Catalog No. 89-005-800


Add to cart

Description

Description

The enzyme mevalonate pyrophosphate decarboxylase catalyzes the conversion of mevalonate pyrophosphate into isopentenyl pyrophosphate in one of the early steps in cholesterol biosynthesis. It decarboxylates and dehydrates its substrate while hydrolyzing ATP. [provided by RefSeq

Sequence: MASEKPLAAVTCTAPVNIAVIKYWGKRDEELVLPINSSLSVTLHQDQLKTTTTAVISKDFTEDRIWLNGREEDVGQPRLQACLREIRCLARKRRNSRDGDPLPSSLSCKVHVASVNNFPTAAGLASSAAGYACLAYTLARVYGVESDLSEVARRGSGSACRSLYGGFVEWQMGEQADGKDSIARQVAPESHWPELRVLILVVSAEKKLTGSTVGMRASVETSPLLRFRAESVVPARMAEMARCIRERDFPSFAQLTMKDSNQFHATCLDTFPPISYLNAISWRIIHLVHRFNAHHGDTKVAYTFDAGPNAVIFTLDDTVAEFVAAVWHGFPPGSNGDTFLKGLQVRPAPLSAELQAALAMEPTPGGVKYIIVTQVGPGPQILDDPCAHLLGPDGLPKPAA
Specifications

Specifications

mevalonate (diphospho) decarboxylase
Unconjugated
No additive
NM_002461.1
FP17780|MPD
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
4597
Western Blot
Mouse polyclonal antibody raised against a full-length human MVD protein.
MVD
Serum
MVD
MVD (NP_002452.1, 1 a.a. ~ 400 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit