Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MVD Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA513676

Catalog No. PIPA513676

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The enzyme mevalonate pyrophosphate decarboxylase catalyzes the conversion of mevalonate pyrophosphate into isopentenyl pyrophosphate in one of the early steps in cholesterol biosynthesis. It decarboxylates and dehydrates its substrate while hydrolyzing ATP.
Specifications

Specifications

MVD
Polyclonal
PBS with 0.09% sodium azide
P53602
Diphosphomevalonate decarboxylase; Mevalonate pyrophosphate decarboxylase; Mevalonate (diphospho)decarboxylase; MPD; MVD
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4597
Immunohistochemistry, Western Blot
Unconjugated
MVD
Liquid
MVD
KLH conjugated synthetic peptide between 7-34 amino acids from the N-terminal region of human MVD
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit