Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAX dimerization protein 4, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human MXD4 protein.

Manufacturer:  Abnova Corporation H00010608B02

Catalog No. 89-007-020


Add to cart

Description

Description

This gene is a member of the MAD gene family . The MAD genes encode basic helix-loop-helix-leucine zipper proteins that heterodimerize with MAX protein, forming a transcriptional repression complex. The MAD proteins compete for MAX binding with MYC, which heterodimerizes with MAX forming a transcriptional activation complex. Studies in rodents suggest that the MAD genes are tumor suppressors and contribute to the regulation of cell growth in differentiating tissues. [provided by RefSeq

Sequence: MELNSLLILLEAAEYLERRDREAEHGYASVLPFDGDFAREKTKAAGLVRKAPNNRSSHNELEKHRRAKLRLYLEQLKQLVPLGPDSTRHTTLSLLKRAKVHIKKLEEQDRRALSIKEQLQQEHRFLKRRLEQLSVQSVERVRTDSTGSAVSTDDSEQEVDIEGMEFGPGELDSVGSSSDADDHYSLQSGTGGDSGFGPHCRRLGRPALS
Specifications

Specifications

MAX dimerization protein 4
Unconjugated
No additive
NM_006454.2
MAD4|MST149|MSTP149|bHLHc12
Mouse
50 uL
Cell Cycle
Y
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human MXD4 protein.
MXD4
Serum
MXD4
MXD4 (NP_006445.1, 1 a.a. ~ 209 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
10608
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit