Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYBPC2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521694

Catalog No. PIPA521694


Add to cart

Description

Description

PA5-21694 targets MYBPC2 in IF and WB applications and shows reactivity with Human samples. The PA5-21694 immunogen is a recombit fragment corresponding to a region within amino acids 411 and 729 of MYBPC2 (Uniprot IDNo.Q14324). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

MYBPC2
Polyclonal
Unconjugated
MYBPC2
Liquid
MYBPC2
Recombit fragment corresponding to a region within amino acids 411 and 729 of MYBPC2 (Uniprot IDNo.Q14324)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Western Blot
0.87 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q14324
MYBPC, MYBPCF, C-protein, skeletal muscle fast isoform, fast MyBP-C, fast-type muscle myosin-binding-protein C, myosin-binding protein C, fast-type
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
4606
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit