Learn More
UbpbioSupplier Diversity Partner ANTI-MYC, MOUSE 100UG DFS Item
Manufacturer:  UbpbioSupplier Diversity Partner Y1091
Antibodies – Anti-MYC (UBPBio catalog# Y1091) – Mouse monoclonal antibody (clone MYC.A7, IgG1). Used for immunoblotting at 1:1,000-3,000 dilution for detection of 10-50 ng Myc-tagged proteins. 1:500-2,000 dilution for immunostaining, 1-3 ug Myc antibody/reaction for immunoprecipitation.
Catalog No. 50-112-6924