Learn More
Myc Rabbit anti-Human, Mouse, Avian, Xenopus, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 06340
Catalog No. 06-340-MI
Add to cart
Description
Description
Use Anti-Myc Antibody (rabbit polyclonal antibody) validated in Chimmunoprecipitation, immunocytochemistry, immunoprecipitation, western blotting to detect Myc also known as Transcription factor p64, avian myelocytomatosis viral oncogene homolog, myc proto-oncogene protein.
Specifications
Specifications
Myc
Unconjugated
Purified
Rabbit
IgG
100μg
Primary
NP_002458
Immunocytochemistry, Immunoprecipitation, Western Blot
Purified rabbit polyclonal IgG in buffer containing PBS, 0.05% sodium azide with 2mg/mL BSA. Frozen solution.
MYC; C-MYC; c-Myc
Bacterially expressed fusion-protein corresponding to the full-length human Myc.
Immunoaffinity chromatography
−20°C in undiluted aliquotes for up to 12 months
Polyclonal
Avian, Human, Murine, Xenopus