Learn More
Myc Tag Mouse anti-Human, Clone: 4A6, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05724MG
Catalog No. 05-724-MGMI
Add to cart
Description
Description
Detect Myc Tag using this Anti-Myc Tag Antibody, clone 4A6 validated for use in Chimmunoprecipitation, immunofluoresence, immunoprecipitation, western blotting and immunocytochemistry.
Specifications
Specifications
Myc Tag
4A6
PBS
Purified
Murine
IgG1
1mg
Primary
Human
Chromatin Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
NM_002467.3
MYC, C-MYC, c-Myc
peptide corresponding to amino acids 410-420 (MEQKLISEEDL) of human Myc
Protein G purfied
−20°C for 24 months.
Monoclonal