Learn More
Myc Tag Mouse anti-Human, Clone: 4A6, MilliporeSigma™ Available on GSA/VA Contract for Federal Government customers only.
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05724
Catalog No. 05-724-MI
Add to cart
Description
Description
This Anti-Myc Tag Antibody, clone 4A6 is validated for use in Chimmunoprecipitation, immunocytochemistry, immunofluoresence, immunoprecipitation, western blotting for the detection of Myc Tag. This antibody has been cited in more than 25 published papers.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Myc Tag
4A6
Protein A Purified immunoglobulin in 0.1M Tris-Glycine, 0.15M NaCl, 0.05 Sodium Azide, pH 7.4.
Purified
Murine
IgG
200μg
Primary
Human
Chromatin Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
NM_002467.3
MYC, C-MYC, c-Myc
peptide corresponding to amino acids 410-420 (MEQKLISEEDL) of human Myc
Protein G purfied
2-8°C in undiluted aliquots for up to 12 months.
Monoclonal