Learn More
Myc Tag Mouse anti-All, Clone: 9E10, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05419
Catalog No. 05-419-MI
Add to cart
Description
Description
Anti-Myc Tag Antibody, clone 9E10 is an antibody against Myc Tag for use in immunohistochemistry (paraffin), immunocytochemistry, immunoprecipitation and western blotting.
Specifications
Specifications
Myc Tag
9E10
Protein G Purified immunoglobulin in 0.07M Tris-glycine, pH 7.4, 0.105MNaCl, 0.035% sodium azide as a preservative.
Purified
Murine
IgG1
200μg
Primary
All
Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Unconjugated
NM_002467.3
MYC, C-MYC, c-Myc
Synthetic peptide corresponding to amino acids 408-437 of the Leucine zipper region of human Myc (AEEQKLISEEDLLRKRREQLKHKLEQLRNS).
Protein G purfied
−20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles.
Monoclonal