Learn More
Myc Tag Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 06549
Catalog No. 06-549-MI
Add to cart
Description
Description
Anti-Myc Tag Antibody is an antibody against Myc Tag for use in IC and western blotting.
Specifications
Specifications
Myc Tag
Unconjugated
Purified
Rabbit
IgG
200μg
Primary
NM_002467.3
Immunocytochemistry, Western Blot
Purified rabbit in buffer containing 0.1M Tris-glycine, pH 7.4, 0.15M NaCl with 0.05% sodium azide. Frozen solution.
MYC; C-MYC; c-Myc
Synthetic peptide corresponding to residues 409-420 of human Myc (MEQKLISEEDLN).
Protein A chromatography
−20°C for up to 12 months
Polyclonal
Human
Documents
Promotions
Promotions
  • MilliporeSigma
    Buy two qualifying MilliporeSigma antibodies and get a third of equal or lesser value at no additional cost.
    12-31-2020