Learn More
Mycobacterium avium Mouse anti-Bacteria, Clone: JD11-366.1, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA110789
Catalog No. 01-671-819
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
MA1-10789 detects Mycobacterim avium. MA1-10789 has been successfully used in ELISA applications. The MA1-10789 immunogen is M. avium cell extract.
Mycobacteria are straight or slightly curved, non motile rods that cause TB. Mycobacteria avium causes TB in birds and a pulmonary infection of humans. It can also be transmitted to immunocompromised humans such as AIDS patients where it can cause disseminated Mycobacterium avium complex (MAC).
Specifications
Specifications
Mycobacterium avium
Monoclonal
∽1 mg/mL
PBS with no preservative; pH 7.4
M. avium
M. avium cell extract.
Purified
RUO
Antibody
Monoclonal
ELISA
JD11-366.1
Unconjugated
Liquid
Mouse
IgG1
500 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
Bacteria