Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MyD88 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA141072

Catalog No. 01-676-667


Add to cart

Description

Description

Suggested positive control: human spleen or RAW.

MyD88, a protein involved in interleukin-1 (IL-1) mediated signalling was originally isolated as myeloid differentiation primary response gene (3,4). MyD88 possesses a N-terminal death domain similar to cytoplasmic segments of TNF receptor 1, Fas, and C-terminal region related to IL-1 and Toll receptors. Overexpression of MyD88 induces activation the c-Jun N-terminal kinase and NF-kB through its death domain (1,2). The C-terminus of MyD88 interacts with the IL-1 receptor and blocks NF-kB activation induced by IL-1, but not by NF-kB.
Specifications

Specifications

MyD88
Polyclonal
Unconjugated
MYD88
Liquid
MYD88
Synthetic peptide corresponding to residues 233-248 of human MyD88.
Protein G
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 0.05% sodium azide
Q99836, P22366, Q6Y1S1
MYD88D
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
17874, 301059, 4615
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit