Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myeloid-Associated Differentiation Marker Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 250-300 of human MYADM was used as the immunogen for this antibody.
Antigen Myeloid-Associated Differentiation Marker
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224494SS
View Documents
Novus Biologicals
NBP224494SS
0.025 mg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
NBP224494
View Documents
Novus Biologicals
NBP224494
0.1 mg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Myeloid-Associated Differentiation Marker Polyclonal antibody specifically detects Myeloid-Associated Differentiation Marker in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin).
Specifications

Specifications

Rabbit
A portion of amino acids 250-300 of human MYADM was used as the immunogen for this antibody.
Polyclonal
Primary
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)
Q96S97
PBS with 0.05% Sodium Azide
IgG
Myeloid-Associated Differentiation Marker
Affinity Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a and CRA_b), horse, rat (isoform CRA_a), mouse (isoforms CRA_b and isoform CRA_d), chimpanzee, dog and rhesus monkey, 93% homologous in opossum and 86% homologous in zebrafish.
91663
1.0 mg/mL
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit