Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma anti-Myeloperoxidase, Polyclonal
Rabbit Polyclonal Antibody
Manufacturer: MilliporeSigma 4759151ML
Description
Specifically detects Myeloperoxidase Clone: in Human samples, and it is validated for ELISA, Immunoelectrophoresis
Specifications
Myeloperoxidase | |
Unconjugated | |
purified | |
human neutrophil myeloperoxidase | |
1mL | |
−20°C | |
Polyclonal | |
Human |
ELISA, Immunoelectrophoresis | |
In PBS. | |
Rabbit | |
IgG | |
RUO | |
Primary | |
Recognizes myeloperoxidase in human neutrophil granules. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only