Learn More
Myeloperoxidase Rabbit anti-Human, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA122870
Catalog No. PA122870
Add to cart
Description
Description
PA1-22870 detects myeloperoxidase in human and rat samples. PA1-22870 has been successfully used in immunofluorescence, and immunohistochemistry (frozen and paraffin) procedures. PA1-22870 immunogen is myeloperoxidase isolated from human granulocytes.
Myeloperoxidase (MPO) is a hemoprotein that is abundantly expressed in neutrophils and secreted during their activation. Native Myeloperoxidase is represented as a covalently bound tetrameric complex of two glycosylated alpha chains with a molecular weight range of 59 - 64 kDa, and two unglycosylated beta chains (MW ~14 kDa) with total MW ~150 kDa and a theoretical pI of 9.2. Traditionally, myeloperoxidase was considered as a main target of anti-neutrophil cytoplasm antibodies (ANCA), the serological markers for certain systemic vasculities such as periarteriitis nodosa, microscopic polyarteriitis and pulmonary eosinophilic granulomatosis (Churg-Strauss syndrome). Low to moderate anti-myeloperoxidase autoantibody levels are also reported in rheumatoid arthritis. Recently, it was shown that myeloperoxidase participates in the initiation and progression of cardiovascular disease. Myeloperoxidase possesses potent proinflammatory properties and may contribute directly to tissue injury. Now Myeloperoxidase is under consideration as one of the most promising cardiac markers. Myeloperoxidase is part of the host defense system of polymorphonuclear leukocytes and myeloperoxidase is responsible for microbicidal activity against a wide range of organisms. In the stimulated PMN (polymorphonuclear leukocytes), MPO catalyzes the production of hypohalous acids, primarily hypochlorous acid in physiologic situations, and other toxic intermediates that greatly enhance PMN microbicidal activity.
Specifications
Specifications
Myeloperoxidase
Polyclonal
Unconjugated
MPO
Liquid
MPO
Myeloperoxidase isolated from human granulocytes.
Protein A
RUO
Antibody
Polyclonal
Human, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
0.5 mg/mL
PBS with 1% BSA and 0.05% sodium azide; pH 7.5
P05164
MPO
Rabbit
IgG
500 μL
4° C, do not freeze
Primary
303413, 4353