Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYF6 Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP152030
Description
MYF6 Polyclonal specifically detects MYF6 in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
MYF6 | |
Polyclonal | |
Western Blot 0.5 - 1 ug/ml, Peptide ELISA Detection limit 1:32000 | |
NP_002460.1 | |
MYF6 | |
Peptide with sequence C-SYRPKQENLEGAD corresponding to internal region according to NP_002460.1. | |
0.1 mg | |
Primary | |
Cross reacts with Human. Expected from sequence similarity: Mouse, Rat and Dog. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
bHLHc4BHLHC4, Class C basic helix-loop-helix protein 4, MRF4, MRF4myogenic factor 6, Muscle-specific regulatory factor 4, Myf-6, myogenic factor 6 (herculin) | |
Goat | |
Affinity Purified | |
RUO | |
4618 | |
Human | |
IgG |
missing translation for 'provideContentCorrection'
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'
For Research Use Only
Spot an opportunity for improvement?
missing translation for 'provideContentCorrection'