Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-MYH, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 7 publications

Manufacturer:  Novus Biologicals NB6001032

 View more versions of this product

Catalog No. NB6001032


Add to cart

Description

Description

MYH Polyclonal specifically detects MYH in Human, Mouse, Rat, Bacteria samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

MYH
0.5 mg/mL
Western Blot 2 ug/ml, Immunohistochemistry 1:10- 1:2000, Immunocytochemistry/Immunofluorescence 18 ug/ml
NP_001121897
CYP2C, EC 3.2.2, EC 3.2.2.-, hMYH, MGC4416, mutY (E. coli) homolog, MutY homolog, mutY homolog (E. coli), Muyth, MYHA/G-specific adenine DNA glycosylase
Rabbit
IgG
0.1 mg
Base Excision Repair, Cancer, DNA Repair
Primary
4595
Human, Mouse, Rat, Bacteria
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Unconjugated
PBS containing 0.05% BSA with 0.02% Sodium Azide
Purified
MUTYH
This antibody was generated by immunizing rabbits with a synthetic peptide corresponding to amino acids 531-546 of human MYH.
Protein G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Cross-reacts with Human, Mouse, Rat and Bacteria. Not yet tested in other species.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only