Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYH9 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527506

Catalog No. PIPA527506


Add to cart

Description

Description

PA5-27506 targets MYH9 in IHC (P) and WB applications and shows reactivity with Human samples. The PA5-27506 immunogen is recombit fragment corresponding to a region within amino acids 1 and 184 of Human MYH9. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene encodes a myosin IIA heavy chain that contains an IQ domain and a myosin head-like domain. The protein is involved in several important functions, including cytokinesis, cell motility and maintece of cell shape. Defects in MYH9 are the cause of non-syndromic sensorineural deafness autosomal domit type 17, Epstein syndrome, Alport syndrome with macrothrombocytopenia, Sebastian syndrome, Fechtner syndrome and macrothrombocytopenia with progressive sensorineural deafness.
Specifications

Specifications

MYH9
Polyclonal
Unconjugated
MYH9
Liquid
MYH9
Recombit fragment corresponding to a region within amino acids 1 and 184 of Human MYH9
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
0.55 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
P35579
BDPLT6, DFNA17, EPSTS, FTNS, MHA, NMHC-II-A, NMMHC-IIA, NMMHCA, cellular myosin heavy chain, type A, myosin heavy chain 9, myosin heavy chain, non-muscle IIa, myosin-9, non-muscle myosin heavy chain A, non-muscle myosin heavy chain IIa, non-muscle myosin heavy polypeptide 9, nonmuscle myosin heavy chain II-A
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
4627
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit