Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYLK4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525869

Catalog No. PIPA525869


Add to cart

Specifications

Specifications

MYLK4
Polyclonal
PBS with 0.09% sodium azide
Q86YV6
SgK085, caMLCK like, myosin light chain kinase family member 4, sugen kinase 85
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
340156
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
MYLK4
Liquid
MYLK4
KLH conjugated synthetic peptide between 286-314 amino acids from the C-terminal region of human MYLK4
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit