Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYO1B Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA522347

Catalog No. PIPA522347


Add to cart

Description

Description

PA5-22347 targets MYO1B in WB applications and shows reactivity with Human samples. The PA5-22347 immunogen is a recombit fragment corresponding to a region within amino acids 916 and 1136 of MYO1B (Uniprot IDNo.O43795). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

MYO1B
Polyclonal
Unconjugated
MYO1B
Liquid
MYO1B
Recombit fragment corresponding to a region within amino acids 916 and 1136 of MYO1B (Uniprot IDNo.O43795)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
0.91 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
O43795
myr1, MMI-alpha, MMIa, MYH-1c, MYO1B variant protein, myosin I alpha, myosin-I alpha, myosin-Ib
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
4430
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit