Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYOCD Mouse anti-Human, Mouse, Clone: 355521, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524103

Catalog No. PIMA524103


Add to cart

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 µg of the antibody by the LAL method.

Specifications

Specifications

MYOCD
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q8IZQ8, Q8VIM5
BSAC2A, MYCD, MYOCD, Srfcp, MYCD, myocardin
Mouse
IgG2B
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
214384, 93649
Western Blot
355521
Unconjugated
MYOCD
Lyophilized
MYOCD
E. coli-derived recombit human Myocardin Met97-Leu290
Protein A/G
RUO
Antibody
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit