Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Myocilin Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 3 publications
Supplier: R&D Systems MAB3446
This item is not returnable.
View return policy
Description
Myocilin Monoclonal specifically detects Myocilin in Human samples. It is validated for Western Blot.Specifications
Myocilin | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
Q99972 | |
MYOC | |
Mouse myeloma cell line NS0-derived recombinant human Myocilin Arg33-Met504 Accession # Q99972 | |
100 μg | |
Primary | |
Detects human Myocilin in direct ELISAs and Western blots. In direct ELISAs, 75% cross-reactivity with recombinant mouse Myocilin is observed. | |
Human | |
Purified |
Western Blot | |
297817 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
GLC1A, GLC1Amyocilin, GPOA, JOAG, JOAG1, MYOC, myocilin, trabecular meshwork inducible glucocorticoid response, TIGR, TIGRmutated trabecular meshwork-induced glucocorticoid response protein, Trabecular meshwork-induced glucocorticoid response protein | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4653 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction