Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myoglobin Sheep anti-Human, Polyclonal, Invitrogen™

Sheep Polyclonal Antibody

Manufacturer:  Invitrogen PA175298

Catalog No. PIPA175298

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PA1-75298 specifically detects Myoglobin. PA1-75298 has been successfully used in ELISA applications. The PA1-75298 immunogen is Human myoglobin (cardiac).

This gene encodes a member of the globin superfamily and is expressed in skeletal and cardiac muscles. The encoded protein is a haemoprotein contributing to intracellular oxygen storage and transcellular facilitated diffusion of oxygen. At least three alternatively spliced transcript variants encoding the same protein have been reported.
Specifications

Specifications

Myoglobin
Polyclonal
PBS with 0.09% sodium azide
P02144
Human myoglobin
Sheep
IgG
1 mL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4151
ELISA
Unconjugated
MB
Liquid
MB
Human myoglobin (cardiac)
Purified
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit