Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYO5A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA124947
Description
PA1-24947 detects MYO5A from rat and chicken samples. PA1-24947 will not react with mouse. PA1-24947 has been successfully used in Western blot procedures. The PA1-24947 immunogen is a synthetic peptide, conjugated to KLH, corresponding to residues 1782-1799 of chicken MYO5A.
This gene is one of three myosin V heavy-chain genes, belonging to the myosin gene superfamily. Myosin V is a class of actin-based motor proteins involved in cytoplasmic vesicle transport and anchorage, spindle-pole alignment and mRNA translocation. The protein encoded by this gene is abundant in melanocytes and nerve cells. Mutations in this gene cause Griscelli syndrome type-1 (GS1), Griscelli syndrome type-3 (GS3) and neuroectodermal melanolysosomal disease, or Elejalde disease. Multiple alternatively spliced transcript variants encoding different isoforms have been reported, but the full-length nature of some variants has not been determined.Specifications
MYO5A | |
Polyclonal | |
Unconjugated | |
Q02440, Q9QYF3 | |
MYO5A | |
Synthetic peptide, conjugated to KLH, corresponding to residues 1782-1799 of chicken MYO5A. | |
100 μL | |
Primary | |
Chicken, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Myo5a | |
GS1, MYH12, MYO5, MYR12 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25017, 396237 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction