Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYO9B Rabbit anti-Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA124950

Catalog No. PIPA124950


Add to cart

Description

Description

PA1-24950 detects MYO9B from rat samples. PA1-24950 is expected to cross react with mouse and human due to sequence homology. PA1-24950 has been successfully used in Western blot procedures. The PA1-24950 immunogen is a synthetic peptide, conjugated to KLH, corresponding to residues 1960-1980 of rat MYO9B. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

displays ATP-dependent binding to actin: binds members of the rho family of GTPases; may play a role in formation of the actin cytoskeleton.
Specifications

Specifications

MYO9B
Polyclonal
0.01M PBS with 15mM sodium azide; pH 7.4
Q13459, Q9QY06, Q63358
MYO9B
Synthetic peptide, conjugated to KLH, corresponding to residues 1960-1980 of rat MYO9B.
IgG fraction
RUO
Antibody
Polyclonal
Rat
Western Blot
Unconjugated
MYO9B
Liquid
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
17925, 25486, 4650
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit