Learn More
Myosin Heavy Chain Mouse anti-Human, Rat, Mouse, Zebrafish, Drosophila, Rabbit, Clone: A4.1025, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05716
Catalog No. 05-716-MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Anti-Myosin Heavy Chain Antibody, clone A4.1025 is an antibody against Myosin Heavy Chain for use in immunohistochemistry and western blotting.
Specifications
Specifications
Myosin Heavy Chain
A4.1025
Unpurified mouse ascites monoclonal IgG in buffer containing 0.07MTris-glycine, pH 7.4, 0.105MNaCl, 0.035% sodium azide and 30% glycerol. Liquid at −20°C.
Ascites
Murine
IgG
200μL
Primary
Drosophila, Human, Murine, Rabbit, Rat, Zebrafish
Immunohistochemistry, Western Blot
Unconjugated
NM_005963
MYH1, MyHC-2X/D, MGC133384, MYHa, MyHC-IIx/d, MyHC-2x, Myosin-1, MYHSA1
Partially purified protein from adult human skeletal muscle myosin. Clone A4.1025.
Unpurified
−20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Monoclonal