Learn More
anti-Myosin light chain 3, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP133303
Catalog No. NBP133303
Add to cart
Description
Description
Myosin light chain 3 Polyclonal antibody specifically detects Myosin light chain 3 in Human, Mouse samples. It is validated for Western Blot.
Specifications
Specifications
Myosin light chain 3
0.52mg/mL
Western Blot 1:1000-1:10000
Affinity Purified
MYL3
Recombinant protein encompassing a sequence within the center region of human Myosin Light Chain 3. The exact sequence is proprietary.
22 kDa
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Not yet tested in any other species.
Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Cardiac myosin light chain 1, CMH8Ventricular/slow twitch myosin alkali light chain, CMLC1, light polypeptide 3, alkali; ventricular, skeletal, slow, MLC1SBMyosin light chain 1, slow-twitch muscle B/ventricular isoform, MLC1V, myosin light chain 3, myosin, light chain 3, alkali; ventricular, skeletal, slow, VLC1
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
4634
Human, Mouse