Learn More
MilliporeSigma anti-Myosin, Polyclonal
Rabbit Polyclonal Antibody
Manufacturer: MilliporeSigma 476126500UL
Description
Specifically detects Myosin Clone: in Guinea Pig, Human, Mouse, Rat samples, and it is validated for Immunofluorescence, Immunohistochemistry (Frozen), Immunoprecipitation, Immuno Blotting
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Myosin | |
Unconjugated | |
Undiluted Serum | |
guinea pig muscle myosin | |
500μL | |
≤ -70°C | |
Polyclonal | |
Guinea Pig, Human, Murine, Rat |
Immuno Blotting, Immunoblot, Immunofluorescence, Immunohistochemistry (Frozen), Immunoprecipitation | |
Undiluted serum. | |
Rabbit | |
IgG | |
RUO | |
Primary | |
Recognizes skeletal muscle myosin. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only