Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myosin Vb/Myr 6 Mouse anti-Rat, Unlabeled, Clone: 18, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611608

Catalog No. BDB611608


Add to cart

Description

Description

Myosins are proteins that form oligomers consisting of two heavy chains and varying numbers of light chains. Many classes of myosin superfamily proteins which have similar N-terminal motor domains but divergent C-terminal domains have been characterized. Class V myosins interact via their C-terminal tails with specific proteins involved in organelle transport. In vertebrates, the myosin V family includes mouse myosin Va (dilute myosin) and Vb, rat myosin Vb (myosin from rat 6; myr 6), and chicken p190. Myr 6 includes an N-terminal globular domain, six IQ motifs in the neck region, two coiled-coil domains on each side (cc) of a PEST calpain cleavage region, and a C-terminal tail that is homologous to dilute myosin, AF-6, and GAD. Myr 6 is expressed highly in mouse heart, kidney, testis, liver, and lung. In rat brain, it is expressed highly in dentate gyrus,medial mamillary nuclei, amygdala, and choroid plexus. The tail region of myr 6 interacts with the C-terminal domain of BERP, a RING-B-box-coiled-coil protein that has been implicated in PC12 cell neurite outgrowth. Thus, myr 6 may function in the transport of organelles in a variety of neural and non-neuronal tissues.

Host Species: Mouse
Clone: 18
Isotype: IgG1
Species Reactivity [for Features Main]: Rat
Immunogen: Rat Myr6 aa. 940-1051

Immunofluorescence, Western Blotting

Specifications

Specifications

Myosin Vb/Myr 6
18
Unconjugated
Affinity Purified
Rat Myr6 aa. 940-1051
50μg
Cell Biology
Primary
Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit