Learn More
anti-Myosin VB, Polyclonal, Novus Biologicals™
Manufacturer:  Novus BiologicalsSupplier Diversity Partner 46150002
 View more versions of this product
Catalog No. 46-150-002
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Disclaimers:
For Research Use Only
Specifications Description & Specifications
Specifications
Myosin VB
Unlabeled
Q9ULV0
Rabbit
0.05mg
Primary
4645
Human
ELISA,Western Blot
20mM Potassium phosphate, 150mM Sodium Chloride, pH 7.0
Affinity Purified
This antibody is specific for the Middle Region of the target protein.
RUO
Polyclonal
This product is specific for Human MYO5B.