Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Myosin X, Polyclonal, ™ Non-distribution product as customer accommodation.

Manufacturer:  MilliporeSigma™ AB1094/DEL

Catalog No. 50-173-004

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

Myosins are a superfamily of molecular motor proteins found in eukaryotic cells. Motor proteins are involved in muscular contraction, cytokinesis, short-range membrane/vesicle transport and a host of other cellular processes. The myosins are responsible for actin-based motility by using the energy of ATP hydrolysis to generate force and movement along actin filaments. There are many isoforms of myosin of which some have specialized functions in certain cell types (such as muscle), while other isoforms are ubiquitous. Myosin X belongs to a group of myosin isoforms known as unconventional myosins that participate in cellular processes ranging from membrane trafficking to signaling and cell motility. Myosin X is concentrated at the tips of filopodia and Myosin X rich regions travel forward and backward along filopods, (Berg, 2002). It also appears that Myosin X functions in actual filopodia formation, (Bohil, 2006).

Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Mouse, Rat
Epitope: a.a. 936-953
Immunogen: KLH-conjugated synthetic peptide to amino acids 936-953 of human Myosin X.
Molecular Weight: ~108 kDa

Western Blotting

Specifications

Specifications

Myosin X
Rabbit
100μL
Cell Structure
Polyclonal
Unconjugated
KLH-conjugated synthetic peptide to amino acids 936-953 of human Myosin X.
RUO
Primary
Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit