Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTMR4, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521564

Catalog No. PIPA521564

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PA5-21564 targets myotubularin related protein 4 in immunofluorescence, immunohistochemistry (paraffin), and Western blot applications and shows reactivity with Human and mouse samples.The PA5-21564 immunogen is a recombinant fragment corresponding to a region within amino acids 281 and 578 of myotubularin related protein 4 (Uniprot ID#Q9NYA4).
Specifications

Specifications

myotubularin related protein 4
Polyclonal
Unconjugated
MTMR4
Purified
MTMR4
Recombinant fragment corresponding to a region within amino acids 281 and 578 of myotubularin related protein 4 (Uniprot ID#Q9NYA4)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.64mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q9NYA4, Q91XS1
FYVE-DSP2, ZFYVE11, FYVE domain-containing dual specificity protein phosphatase 2, myotubularin-related protein 4, zinc finger FYVE domain-containing protein 11, zinc finger, FYVE domain containing 11
Rabbit
IgG
100μL
Store at -20°C, Avoid Freeze/Thaw Cycles
Primary
9110
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit