Learn More
N-acetylprocainamide Mouse anti-Chemical, Clone: NAPA12-322.4.1, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA110778
Catalog No. 01-671-808
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
MA1-10778 detects N-acetylprocainamide. MA1-10778 has been successfully used in ELISA applications. The MA1-10778 immunogen is N-Acetylprocainamide, conjugated to KLH.
N-Acetylprocainamide (NAPA or acecainide) is the N-acetylated metabolite of procainamide. It is a Class III antiarrhythmic agent, whereas procainamide is a Class Ia antiarrhythmic drug. It is only partially as active as procainamide and may cause cardiac toxicity and kidney failure. [National Center for Biotechnology Information. PubChem Compound Database; CID=4342]
Specifications
Specifications
N-acetylprocainamide
Monoclonal
∽1 mg/mL
PBS with no preservative; pH 7.4
Acecainide; Acekainid; Acecainida; Acecainidum; N-Acetyloprokainamid; C15H23N3O2
N-Acetylprocainamide, conjugated to KLH.
Purified
RUO
Antibody
Monoclonal
ELISA
NAPA12-322.4.1
Unconjugated
Liquid
Mouse
IgG1
500 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
Chemical