Learn More
N-Cadherin Mouse anti-Human, Clone: 13A9, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05915
Catalog No. 05-915-MI
Add to cart
Description
Description
Detect N-cadherin using this Anti-N-Cadherin Antibody, clone 13A9 validated for use in Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, and Western Blotting.
Specifications
Specifications
N-Cadherin
13A9
Immunoaffinity Purified immunoglobulin in 0.02Mphosphate buffer, pH 7.6, 0.25M NaCl, 0.1% sodium azide and 30% glycerol.
Supernatant
Murine
Unpurified
−20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles.
Monoclonal
Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Unconjugated
NM_001792
CDH2, CDw325, CDHN, NCAD, N-cadherin, Neural-cadherin
Bacterially expressed MBP fusion protein to human N-cadherin cytoplasmic domain. Clone 13A9
100μL
Primary
Human