Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SGSH Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521375

Catalog No. PIPA521375


Add to cart

Description

Description

PA5-21375 targets N-sulfoglucosamine sulfohydrolase precursor in WB applications and shows reactivity with Human samples. The PA5-21375 immunogen is a recombit fragment corresponding to a region within amino acids 44 and 230 of SGSH (Uniprot IDNo.P51688). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

SGSH
Polyclonal
Unconjugated
SGSH
Liquid
SGSH
Recombit fragment corresponding to a region within amino acids 44 and 230 of SGSH (Uniprot IDNo.P51688)
Protein A
RUO
Antibody
Polyclonal
Human
Western Blot
2 mg/mL
PBS with 40% glycerol and 0.01% thimerosal; pH 7
P51688
HSS, MPS3A, SFMD, N-sulphoglucosamine sulphohydrolase, heparan sulfate sulfatase, mucopolysaccharidosis type IIIA, sulfoglucosamine sulfamidase, sulphamidase
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
6448
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit