Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Na+/K+ ATPase α-1 Mouse anti-Canine, Human, Mouse, Monkey, Pig, Rat, Rabbit, Sheep, Xenopus, AlexaFluor™555, Clone: C464.6, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05369X555
Description
Detect Na+/K+ ATPase α-1 using this Anti-Na+/K+ ATPase α-1 Antibody, clone C464.6, Alexa Fluor 555 validated for use in flow cytometry, immunofluorescence and western blotting.Specifications
Na+/K+ ATPase α-1 | |
Monoclonal | |
Alexa Fluor 555 | |
NM_152296.3 | |
Mouse | |
Protein G purfied | |
Primary | |
4°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
IgG1 |
Flow Cytometry, Immunofluorescence, Western Blot | |
C464.6 | |
25μg Alexa Fluor™ 555-conjugated protein G purified mouse IgG1κ in 100μL of PBS containing 1% BSA, 0.05% Tween™-20, 0.05% sodium azide. Liquid at 4°C. | |
ATP1A3, MGC13276, RDP, IIIsubunit)., DYT12, +K, Na | |
Purified Na+/K+ ATPase isolated from membrane fractions of rabbit kidney outer medulla. | |
25 μg | |
Canine, Human, Monkey, Mouse, Porcine, Rabbit, Rat, Sheep, Xenopus species | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only